missing translation for 'onlineSavingsMsg'
Learn More

Ismatec™ 2-Stop Tubing, Tygon 3350 SI

Special silicone-based tubing that undergoes a special treatment with platinum to ensure a very smooth internal surface.

Brand:  Ismatec™ SC0632A

 View more versions of this product

Product Code. 10685673

  • 112.00 € / Pack of 6
Estimated Shipment: 09-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Platinum-cured silicone tubing.
  • Ultra smooth inner bore.
  • Biocompatible for life science applications.
  • Low level of protein binding.
  • Entirely non-toxic, non-hemolytic, and non-pyrogenic.
  • Weather, ozone, sunlight, and radiation resistant.
  • Resistant to fungus.
  • Odorless.
Specifications

Specifications

US Pharmacopoea XXIII Cl.Vl, FDA 21 CFR, Part 177.2600. Also exceeds 3A sanitary standards
Autoclavable
Tygon SI Silicone 3350
Pumps that incorporate cassettes into the pumphead design. Life science applications.
6/Pk
I.D. 2.29 mm
200°C
Purple-Black
2.29 mm
400 mm
−60°C
Acids: Limited, Alkaline solutions: Limited, Solvents: Limited, Pressure: Not rec., Vacuum: Good, Viscous media: Fair, Sterile media: Excellent
Autoclavable with steam, 30 minutes at 1 bar (15 psi) and 121°C (250°F), gas sterilization with ethylene oxide, sterilization with radiation up to 2.5 mrad.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ismatec™ 2-Stop Tubing, Tygon 3350 SI > I.D. 2.29 mm, Purple-Black

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.