missing translation for 'onlineSavingsMsg'
Learn More

2-Sulfobenzoic anhydride, 94%, Thermo Scientific Chemicals

Product Code. 11420913
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-08-3
Molecular Formula C7H4O4S
Molecular Weight (g/mol) 184.165
MDL Number MFCD00005879
InChI Key NCYNKWQXFADUOZ-UHFFFAOYSA-N
Synonym 2-sulfobenzoic anhydride, 2-sulfobenzoic acid cyclic anhydride, sulfobenzoic anhydride, o-sulfobenzoic acid anhydride, 3h-2,1-benzoxathiol-3-one, 1,1-dioxide, 3h-benzo c 1,2 oxathiol-3-one 1,1-dioxide, unii-0swh68iq5w, o-sulfobenzoic acid, cyclic anhydride, 2,1-benzoxathiol-3-one-1,1-dioxide, 0swh68iq5w
PubChem CID 65729
IUPAC Name 1,1-dioxo-2,1$l^{6}-benzoxathiol-3-one
SMILES C1=CC=C2C(=C1)C(=O)OS2(=O)=O

Specifications

Melting Point ∼120°C
Boiling Point 184°C to 186°C (18 mmHg)
Quantity 100 g
Beilstein 139893
Sensitivity Moisture sensitive
Formula Weight 184.17
Percent Purity 94%
Chemical Name or Material 2-Sulfobenzoic anhydride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.