missing translation for 'onlineSavingsMsg'
Learn More

2-Thiazolecarboxaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 11976771
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10200-59-6
InChI Key ZGTFNNUASMWGTM-UHFFFAOYSA-N
Synonym 2-thiazolecarboxaldehyde, thiazole-2-carbaldehyde, 2-formylthiazole, thiazole-2-carboxaldehyde, 2-formyl thiazole, thiazolecarboxaldehyde, 2-formyl-1,3-thiazole, 1,3-thiazole-2-carboxaldehyde, 2-thiazolecarbaldehyde, pubchem9929
PubChem CID 2734903
ChEBI CHEBI:43623
IUPAC Name 1,3-thiazole-2-carbaldehyde
SMILES C1=CSC(=N1)C=O

Specifications

Color Yellow to Brown
Density 1.2880g/mL
Boiling Point 76.0°C to 78.0°C (24.0 mmHg)
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C4H3NOS
Refractive Index 1.5735 to 1.5755
Quantity 5 g
Specific Gravity 1.288
Solubility Information Solubility in water: soluble.
Molecular Weight (g/mol) 113.14
Formula Weight 113.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Thiazolecarboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-Thiazolecarboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'