missing translation for 'onlineSavingsMsg'
Learn More

2-Thioxanthine, 98+%, Thermo Scientific Chemicals

Product Code. 11457224
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Thioxanthine used to inhibit MPO (Myeloperoxidase).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Thioxanthine used to inhibit MPO (Myeloperoxidase).

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2487-40-3
Molecular Formula C5H4N4OS
Molecular Weight (g/mol) 168.174
MDL Number MFCD00031505
InChI Key XNHFAGRBSMMFKL-UHFFFAOYSA-N
Synonym 2-thioxanthine, 6-hydroxy-2-mercaptopurine, 2-mercapto-9h-purin-6-ol, 2-thioxanthene, 2-mercapto-6-hydroxypurine, 2-thio-6-oxypurine, 6-hydroxypurine-2-thiol, 2-thioxo-2,3-dihydro-1h-purin-6 7h-one, xanthine, 2-thio, 6h-purin-6-one, 1,2,3,7-tetrahydro-2-thioxo
PubChem CID 1268185
IUPAC Name 2-sulfanylidene-3,7-dihydropurin-6-one
SMILES C1=NC2=C(N1)C(=O)NC(=S)N2

Specifications

Melting Point >300°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 168.17
Percent Purity ≥98%
Chemical Name or Material 2-Thioxanthine
compliance-icons
Product Identifier
  • 2-Thioxanthine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2-Thioxanthine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.