missing translation for 'onlineSavingsMsg'
Learn More

2-(Tri-n-butylstannyl)furan, 97%

Product Code. 15451448
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
This item is not returnable. View return policy

Product Code. 15451448

Brand: Thermo Scientific Alfa Aesar H55259.AA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 118486-94-5 | C16H30OSn | 357.125 g/mol

It is an agrochemical, organic and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an agrochemical, organic and pharmaceutical intermediates.

Notes
Store at room temperature. Light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118486-94-5
Molecular Formula C16H30OSn
Molecular Weight (g/mol) 357.125
MDL Number MFCD00192512
InChI Key SANWDQJIWZEKOD-UHFFFAOYSA-N
Synonym 2-tributylstannyl furan, tributyl furan-2-yl stannane, 2-tributylstannylfuran, tributyl 2-furyl tin, tributyl 2-furyl stannane, 2-tri-n-butylstannyl furan, stannane,tributyl-2-furanyl, stannane, tributyl-2-furanyl, 2-furyltributyltin, tributylstannylfuran
PubChem CID 4393688
IUPAC Name tributyl(furan-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CO1

Specifications

Color Colorless to Yellow
Density 1.134 g/mL
Boiling Point 90°C (0.1 mmHg)
Refractive Index 1.494
Quantity 5 mL
UN Number UN2788
Formula Weight 357.1
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-(Tri-n-butylstannyl)furan

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.