missing translation for 'onlineSavingsMsg'
Learn More

2-(Tri-n-butylstannyl)oxazole, 95%, Thermo Scientific Chemicals

Product Code. 15432258
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-(Tri-n-butylstannyl)oxazole act as a synthetic building block, which is utilized in Stille coupling

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Tri-n-butylstannyl)oxazole act as a synthetic building block, which is utilized in Stille coupling

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145214-05-7
Molecular Formula C15H29NOSn
Molecular Weight (g/mol) 358.113
MDL Number MFCD06798076
InChI Key YOWGRWHKDCHINP-UHFFFAOYSA-N
Synonym 2-tributylstannyl oxazole, 2-tri-n-butylstannyl oxazole, 2-tributylstannyl-1,3-oxazole, 2-tributylstannanyl-oxazole, 2-tributylstannyloxazole, 2tributylstannyloxazole, acmc-20aom1, 2-tributylstannyl-oxazole, 2-tributylstannanyloxazole
PubChem CID 15486183
IUPAC Name tributyl(1,3-oxazol-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=NC=CO1

Specifications

Color Colorless
Density 1.71 g/mL
Boiling Point 108°C to 110°C (0.2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.493
Quantity 250 mg
UN Number UN2788
Solubility Information Insoluble in water.
Formula Weight 358.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-(Tri-n-butylstannyl)oxazole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'