missing translation for 'onlineSavingsMsg'
Learn More

2-(Trichloroacetyl)pyrrole, 98%, Thermo Scientific™

Product Code. 10883021
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 35302-72-8
Molecular Formula C6H4Cl3NO
Molecular Weight (g/mol) 212.46
MDL Number MFCD00128757
InChI Key BBFDGMDENAEMKF-UHFFFAOYSA-N
Synonym 2-trichloroacetyl pyrrole, 2,2,2-trichloro-1-1h-pyrrol-2-yl ethanone, 2,2,2-trichloro-1-1h-pyrrol-2-yl ethan-1-one, 2-trichloroacetyl-1h-pyrrole, 2-trichloroacetylpyrrole, 2,2,2-trichloro-1-1h-pyrrol-2-yl-ethanone, ethanone, 2,2,2-trichloro-1-1h-pyrrol-2-yl, 2,2,2-trichloro-1-pyrrol-2-ylethan-1-one, trichloroacetyl pyrrole
PubChem CID 321487
IUPAC Name 2,2,2-trichloro-1-(1H-pyrrol-2-yl)ethanone
SMILES C1=CNC(=C1)C(=O)C(Cl)(Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 72.0°C to 74.0°C
Color Green
Elemental Analysis 33.07 to 34.76% C, 6.43 to 7.75% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 10 g
Formula Weight 212.46
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trichloroacetyl)pyrrole
Show More Show Less
compliance-icons
Product Identifier
  • 2-(Trichloroacetyl)pyrrole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'