missing translation for 'onlineSavingsMsg'
Learn More
2-Trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 99%, Thermo Scientific Chemicals Product Code.: 11373157

2-Trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 99%, Thermo Scientific Chemicals

Product Code. 11373157
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11373157

Brand: Thermo Scientific Alfa Aesar L13587.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74291-57-9
Molecular Formula C11H9ClF3NO3S
Molecular Weight (g/mol) 327.70
MDL Number MFCD01075651
InChI Key SXIYEBVAQTUBOQ-UHFFFAOYSA-N
Synonym 2-2,2,2-trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 1,2,3,4-tetrahydro-2-trifluoroacetyl isoquinoline-7-sulfonyl chloride, 2-trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 2-2,2,2-trifluoroacetyl-3,4-dihydro-1h-isoquinoline-7-sulfonyl chloride, 1-7-chlorosulfonyl 2-1,2,3,4-tetrahydroisoquinolyl-2,2,2-trifluoroethan-1-one, 2-trifluoroacetyl-7-chlorosulfonyl-1,2,3,4-tetrahydroisoquinoline, 7-chlorosulfonyl-2-trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline, 1,2,3,4-tetrahydro-2-trifluoroacetyl isoquinoline-7-sulfoneShow MoreShow Less
PubChem CID 11324902
IUPAC Name 2-(2,2,2-trifluoroacetyl)-3,4-dihydro-1H-isoquinoline-7-sulfonyl chloride
SMILES FC(F)(F)C(=O)N1CCC2=C(C1)C=C(C=C2)S(Cl)(=O)=O

Specifications

Melting Point 102°C to 106°C
Quantity 1 g
UN Number UN3261
Beilstein 5603124
Formula Weight 327.71
Percent Purity 99%
Chemical Name or Material 2-Trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberFA020658 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL13587.03,L13587.06
Lot Number10152065 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL13587.03,L13587.06
Lot NumberGBFA021692 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL13587.03,L13587.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.