missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)benzaldehyde, 96%, Thermo Scientific™

Product Code. 10722942
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 94651-33-9
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.12
MDL Number MFCD00042405
InChI Key CPHXLFKIUVVIOQ-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy benzaldehyde, 4-trifluoromethoxy, o-trifluoromethoxy benzaldehyde, 2-trifluoromethoxy-benzaldehyde, pubchem1469, trifluoromethoxybenzaldehyde, acmc-209rt0, o-trifluoromethoxybenzaldehyde, trifluoromethoxy benzaldehyde
PubChem CID 2777192
IUPAC Name 2-(trifluoromethoxy)benzaldehyde
SMILES FC(F)(F)OC1=CC=CC=C1C=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 77°C (20.0 mmHg)
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 190.12
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethoxy)benzaldehyde
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.