missing translation for 'onlineSavingsMsg'
Learn More

2'-(Trifluoromethyl)acetophenone, 97+%, Thermo Scientific™

28.00 € - 192.00 €

Chemical Identifiers

CAS 17408-14-9
Molecular Formula C9H7F3O
Molecular Weight (g/mol) 188.149
MDL Number MFCD00000378
InChI Key FYDUUODXZQITBF-UHFFFAOYSA-N
Synonym 2'-trifluoromethyl acetophenone, 1-2-trifluoromethyl phenyl ethanone, o-trifluoromethylacetophenone, 1-2-trifluoromethyl phenyl ethan-1-one, 2'-trifluoromethylacetophenone, ethanone, 1-2-trifluoromethyl phenyl, o-trifluoromethyl benzoyl, 2-trifluoromethylacetophenone, 1-acetyl-2-trifluoromethyl benzene
PubChem CID 87095
IUPAC Name 1-[2-(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=CC=CC=C1C(F)(F)F
View More Specs

Products 3
Product Code Brand Quantity Price Menge & Verfügbarkeit  
Produktcode Marke Quantity Preis Menge & Verfügbarkeit  
11404953
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A12248.03
1 g
28.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
11414953
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A12248.06
5 g
45.80 €
5g
Please to purchase this item. Need a web account? Register with us today!
11424953
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A12248.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Beschreibung

Beschreibung

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Spezifikation

Chemische Identifikatoren

17408-14-9
188.149
FYDUUODXZQITBF-UHFFFAOYSA-N
87095
CC(=O)C1=CC=CC=C1C(F)(F)F
C9H7F3O
MFCD00000378
2'-trifluoromethyl acetophenone, 1-2-trifluoromethyl phenyl ethanone, o-trifluoromethylacetophenone, 1-2-trifluoromethyl phenyl ethan-1-one, 2'-trifluoromethylacetophenone, ethanone, 1-2-trifluoromethyl phenyl, o-trifluoromethyl benzoyl, 2-trifluoromethylacetophenone, 1-acetyl-2-trifluoromethyl benzene
1-[2-(trifluoromethyl)phenyl]ethanone

Spezifikation

17408-14-9
1.255
84°C (183°F)
C9H7F3O
MFCD00000378
2'-trifluoromethyl acetophenone, 1-2-trifluoromethyl phenyl ethanone, o-trifluoromethylacetophenone, 1-2-trifluoromethyl phenyl ethan-1-one, 2'-trifluoromethylacetophenone, ethanone, 1-2-trifluoromethyl phenyl, o-trifluoromethyl benzoyl, 2-trifluoromethylacetophenone, 1-acetyl-2-trifluoromethyl benzene
CC(=O)C1=CC=CC=C1C(F)(F)F
188.149
188.15
2'- (Trifluoromethyl)acetophenone
15°C to 17°C
163°C to 165°C
Characteristic
1.4595
2259035
FYDUUODXZQITBF-UHFFFAOYSA-N
1-[2-(trifluoromethyl)phenyl]ethanone
87095
≥97%
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

missing translation for 'einecsNumber' : 241-434-2

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate