missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)thioxanthen-9-one, 98%, Thermo Scientific™

Product Code. 15422229
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1693-28-3
Molecular Formula C14H7F3OS
Molecular Weight (g/mol) 280.26
MDL Number MFCD00079785
InChI Key NEWRXGDGZGIHIS-UHFFFAOYSA-N
Synonym 2-trifluoromethyl thioxanthen-9-one, 2-trifluoromethyl thioxanthone, 2-trifluoromethyl-9h-thioxanthen-9-one, 2-trifluoromethylthioxanthone, 9h-thioxanthen-9-one, 2-trifluoromethyl, 2-trifludromethylthioxanthone, 2-trifluormethyl-9-thiaxanthen-9-one, 2-trifluoromethyl dibenzo b,e thiin-10-one, pubchem10678, acmc-209dzj
PubChem CID 74324
SMILES FC(F)(F)C1=CC=C2SC3=CC=CC=C3C(=O)C2=C1

Specifications

Melting Point 147°C to 151°C
Color Yellow
Quantity 25 g
Beilstein 1583738
Sensitivity Light sensitive
IUPAC Name 2-(trifluoromethyl)thioxanthen-9-one
Formula Weight 280.26
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 2-(Trifluoromethyl)thioxanthen-9-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.