Learn More
Ohaus™ 2 x Centrifuge Rack Adapters, Frontier™ R-S4x100/4MI Swing Out Rotors
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ R-S4x100/4MI Swing Out Rotors
200.00 €
Specifications
For Use With (Equipment) | R-S4x100/4MI Swing Out Rotors |
---|---|
No. per Pack | 2 |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
15816692
|
Ohaus™
30314854 |
10 x 1.5 mL |
200.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15826692
|
Ohaus™
30314855 |
9 x 4.5 mL |
200.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15836692
|
Ohaus™
30314856 |
7 x 5 to 7 mL (13 mm dia., RB) |
200.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15806692
|
Ohaus™
30314853 |
200.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15846692
|
Ohaus™
30314857 |
7 x 5 to 7 mL (13 mm dia., Vac) |
200.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15856692
|
Ohaus™
30314858 |
200.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15896682
|
Ohaus™
30314852 |
200.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15886682
|
Ohaus™
30314851 |
200.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15876682
|
Ohaus™
30314850 |
200.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15866682
|
Ohaus™
30314849 |
200.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15856682
|
Ohaus™
30314848 |
200.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15826682
|
Ohaus™
30314845 |
1 x 50 mL (29 mm dia.,RB) |
200.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15836682
|
Ohaus™
30314846 |
1 x 50 mL (29 mm dia., FA) |
200.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15846682
|
Ohaus™
30314847 |
1 x 50 mL (29 mm dia.) |
200.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15816682
|
Ohaus™
30314844 |
1 x 50 mL (34 mm dia.) |
200.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15806682
|
Ohaus™
30314843 |
1 x 85 mL |
200.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15896672
|
Ohaus™
30314842 |
200.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
- Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
- Suitable for use in Frontier™ R-S4x100/4MI Swing Out Rotors
Specifications
R-S4x100/4MI Swing Out Rotors | |
2 |