missing translation for 'onlineSavingsMsg'
Learn More

Ohaus™ 2 x Centrifuge Tube Adapters, Frontier™ Rotors

Facilitate centrifugation of tubes smaller than the original, provided carrier space for an array of Frontier™ fixed angle rotors.

Brand:  Ohaus™ 30130892

115.50 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15816642

  • 123.00 € / Pack of 2

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Accommodates centrifuge tubes that are smaller than the space provided in the rotor carrier.
  • Suitable for use in the following Frontier™ rotors:
    • R-A12x15/6M
    • R-A6x50/6MI
    • R-A12x15/6MI
    • R-A30x15/4MI
    • R-A10x50/10MI
TRUSTED_SUSTAINABILITY
Specifications

Specifications

16 mL
Frontier Fixed-Angle Rotors
1
18 mm
Adapter, D18mm, 16ml Tubes x2
2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.