missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh

Made from Stainless Steel and available in a wide variesty of sizes. Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh has a 200mm diameter.

Brand:  Fisherbrand™ 01/200SBW3.35

 View more versions of this product

Product Code. 11567532

  • 106.00 € / Each
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket

Description

Description

100 mm and 200 mm diameter test sieves with Stainless Steel frame and woven wire mesh. Supplied with Certificate of Conformity and record card. Additional mesh sizes and diameter sieves available - please contact Customer Service for details.
Specifications

Specifications

1 Each
Sieve
Stainless Steel
200 mm
Woven Wire
3.35 mm
Certificate of Conformity, BS 410: Part 1: 2000, ISO 3310: Part 1: 2000
Certificate of conformity and record card
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers