Learn More
(20R)-Protopanaxadiol, MedChemExpress
MedChemExpress (20R)-Protopanaxadiol is a triterpenoid saponin metabolite of 20(R)-ginsenoside Rg3 in black ginseng. (20R)-Protopanaxadiol exhibits anti-tumor activity and cytotoxicity, and potently inhibits the growth of Helicobacter pylori.
Brand: Medchem Express HY-N2040
This item is not returnable.
View return policy
Description
(20R)-Protopanaxadiol is a triterpenoid saponin metabolite of 20(R)-ginsenoside Rg3 in black ginseng. (20R)-Protopanaxadiol exhibits anti-tumor activity and cytotoxicity, and potently inhibits the growth of Helicobacter pylori.Specifications
(20R)-Protopanaxadiol | |
White | |
C30H52O3 | |
DMSO : 20 mg/mL (43.41 mM; Need ultrasonic) | |
460.73 | |
98.0% | |
Solid |
7755-01-3 | |
Research | |
5 mg | |
C[C@]([C@@](C[C@H]1O)([H])[C@]2(CC[C@@H]3O)C)(CC[C@@]2([H])C3(C)C)[C@]4([C@@]1([H])[C@]([C@](C)(O)CC/C=C(C)/C)([H])CC4)C | |
460.73 | |
Research | |
COVID-19-immunoregulation |