Learn More
Invitrogen™ 20X Bolt™ MES SDS Running Buffer
Optimized for use with Bolt™ Bis-Tris Plus gels, and are available in a variety of formats
Brand: Invitrogen™ B000202
335.57 EUR valid until 2024-12-31
Use promo code "21951" to get your promotional price.
Additional Details : Weight : 5.23800kg
Description
Bolt™ buffers and sample reducing agents are optimized for use with Bolt™ Bis-Tris Plus gels, and are available in a variety of formats. Note: Bolt™ gel run times may vary depending upon the type of running buffer used.About Bolt™ Bis-Tris Plus Gels
Bolt™ Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of your small- to medium-sized proteins under denaturing conditions. Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt™ gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial. Also use Bolt™ gels to obtain optimal results for your day-to-day protein separation needs. Bolt™ Bis-Tris Plus gels come in four gel types and multiple well formats. A Mini Gel Tank is required for use of all Bolt™ gels.
Specifications
Running Buffer | |
Protein electrophoresis & western blotting | |
Liquid | |
5 L |
20X | |
Bolt | |
Bolt | |
Sodiumdodecyl Sulphate 151-21-3 ( 1-5 ) |
For Research Use Only. Not for use in diagnostic procedures.