missing translation for 'onlineSavingsMsg'
Learn More

uvex 2112 One-Fit Disposable Ear Plugs

Characterized by their ergonomic, closed surface design One-Fit ear plugs are constructed of polyurethane foam for optimal comfort, even during long periods of use.

39.25 € - 42.20 €

Specifications

Color Turquoise
ANSI S3.1974 No
Size Regular
Model 2112 One-Fit
Pairs per Package 200
View More Specs

Products 2
Product Code Brand Packaging Type Price Quantity & Availability  
Product Code Brand Packaging Type Price Quantity & Availability  
17290332
View Documents
uvex
2112045
Bag
42.20 €
200 pairs
Estimated Shipment: 24-12-2024
to see stock.
Add to basket
 
17200342
View Documents
uvex
2112046
Refill Bag
39.25 €
200 pairs
Estimated Shipment: 24-12-2024
to see stock.
Add to basket
 
Description

Description

  • Soft, polyurethane foam ensures comfort even during long periods of use
  • Closed surface prevents foreign bodies from entering
  • For use with noise sources up to 110 db(A)
  • Cordless, ergonomic design
Specifications

Specifications

Turquoise
Regular
200
Uncorded
Ear Plugs
No
EN 352-2
No
2112 One-Fit
37 dB
Oval
Single-use
No
Videos
SDS
Documents

Documents

Product Certifications