Learn More
uvex 2112 Xtra-Fit Disposable Ear Plugs
Ergonomic, pre-formed ear plugs expand slowly to adapt to the size and shape of the ear canal, thus minimizing leakage and ensuring optimal protection.
21.10 € - 58.90 €
Specifications
Color Orange
ANSI S3.1974 No
Size Large
Model 2112 Xtra-Fit
Noise Reduction Rating (NRR) 36 dB
View More Specs
Products 3
Product Code Brand Packaging Type Pairs per Package Price Quantity & Availability  
Product Code Brand Packaging Type Pairs per Package Price Quantity & Availability  
17230342
View Documents
uvex
2112093
Polybag 100
21.10 €
100 pairs
Estimated Shipment: 24-12-2024
to see stock.
Add to basket
17210342
View Documents
uvex
2112060
Polybag 200
42.20 €
200 pairs
Estimated Shipment: 24-12-2024
to see stock.
Add to basket
17220342
View Documents
uvex
2112061
Refill Box 250
58.90 €
250 pairs
Estimated Shipment: 24-12-2024
to see stock.
Add to basket
Description
Description
  • Ergonomically, pre-shaped disposable earplugs
  • Slowly expand to adapt to the size and shape of the ear canal
  • Minimized leakage for optimal protection
  • Suitable for larger ear canals
  • Available in hygienic polybags, or refill bags
Specifications
Specifications
Orange
Large
36 dB
Oval
Single-use
No
No
2112 Xtra-Fit
Uncorded
Ear Plugs
No
EN 352-2