missing translation for 'onlineSavingsMsg'
Learn More

uvex 2124 Xact-Fit Multi Reusable Ear Plugs with Storage Case

Innovative, corded ear plugs feature a unique oval design that adjusts perfectly to the anatomy of the ear canal. The angled pin significantly reduces pressure in the ear, while remaining comfortable to wear for long periods of time.

115.00 €

Specifications

Includes Pocket-size case
Color Transparent, Gray
ANSI S3.1974 No
For Use With (Application) Variable Environments
Model 2124 Xact-Fit Multi
View More Specs

Products 2
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17290342
View Documents
uvex
2124017
Small
115.00 €
Pack of 50
Estimated Shipment: 24-12-2024
to see stock.
Add to basket
 
17200352
View Documents
uvex
2124018
Regular
115.00 €
Pack of 50
Estimated Shipment: 24-12-2024
to see stock.
Add to basket
 
Description

Description

  • Unique oval design adjusts perfectly to the anatomy of the ear canal
  • Angled pin significantly reduces pressure in the ear
  • Pre-formed pins make it easy to insert and remove ear plugs
  • Thumb indentations for enable quick and easy handling
  • Smooth, dirt-repellent surface can be cleaned with mild soap and water
  • Comfortably wear corded ear plugs around the neck in low-noise areas
  • Clear, hygienic box included for safe storage when not in use
Specifications

Specifications

Pocket-size case
No
2124 Xact-Fit Multi
Corded
Ear Plugs
No
EN 352-2
Transparent, Gray
Variable Environments
26 dB
Oval
Reusable
No
Videos
SDS
Documents

Documents

Product Certifications