missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 22mm PE, Cap, Transparent, 4.3mm Center hole, 22 x 8.4mm, Assembled septum

Pre-assembled caps and septa are convenient and minimize contamination from handling.

Brand:  Fisherbrand™ 22 15 0321

 View more versions of this product

Product Code. 11869420

  • 316.00 € / Pack of 1000
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • 22mm
  • PE
  • Cap
  • Center hole
  • Combination Seal
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Natural Rubber/TEF
22 mm
Headspace
1.3 mm
Red-Orange, Transparent
Polyethylene
100
Snap Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.