Learn More
Hamilton™ 24 gauge, Large Hub RN Needle, custom length (0.375 to 12 in), point style 2, 3, or 4, 6/PK
Large Hub Removable (RN) needles can be used with removable (RN)-style syringes that are 250 μL and larger. Standard needle length is 2 in. / 51 mm. Removable (RN) needles are available in convenient six packs. Custom lengths and point styles are available.
Brand:  Hamilton™ 7806-06
Product Code. 12396449
  • 197.00 € / Pack of 6
Explore more special offers
Description
Description
Specification Sheet
Gauge24 gauge
Point Style2, 3, or 4
Needle LengthCustom Needle Length
AutoclavableYes
Sales Quantity6 needles per pack
HubLarge RN
Specifications
Specifications
24 G
Large Hub RN