missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 24mm PP Screw Seal, White, Center Hole, 24-400 Thread, Assembled Septum

Ready to use combination seals.

Brand:  Fisherbrand™ 24 15 1394

333.31 EUR valid until 2025-03-14
Use promo code "24142" to get your promotional price.


 View more versions of this product

Product Code. 11530595

  • 446.00 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • No time consuming and tricky assembly
  • No contamination of the liner with sweat/fat normally caused by manual assembly
  • Broad variety of different septa materials for almost all applications
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PTFE/Butyl
24 mm
Storage
2.5 mm
Gray, Red, White
Polypropylene
100
Screw Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.