missing translation for 'onlineSavingsMsg'
Learn More

3M™ 2890 Series Polycarbonate Lens Safety Goggles

19.89 € - 245.00 €

Specifications

Certifications/Compliance EN 166:2001
Lens Tint Clear
No. per Pack 1 Ea.
UV Protection Yes
View More Specs

Products 5
Product Code Brand Lens Coating Ventilation Details Price Quantity & Availability  
Product Code Brand Lens Coating Ventilation Details Price Quantity & Availability  
12672605
View Documents
3M™
7000032483
Anti-fog Sealed
26.00 €
Each
In Stock
to see stock.
Add to basket
 
12642605
View Documents
3M™
7000061924
Anti-fog, Scratch-resistant Sealed
24.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
12652605
View Documents
3M™
7000032481
Anti-fog Indirect ventilation
29.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
10361910
View Documents
3M™
7000032480
Scratch-resistant Indirect ventilation This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
13458997
View Documents
3M™
7000032480
Scratch-resistant Indirect ventilation
245.00 €
Pack of 10
Estimated Shipment: 24-04-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

EN 166:2001
Clear
1 Ea.
Yes
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ 2890 Series Polycarbonate Lens Safety Goggles

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.