missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ 293fectin™ Transfection Reagent

293fectin™ Transfection Reagent is a proprietary, cationic lipid-based formulation for transfecting DNA into eukaryotic cells.

Brand:  Gibco™ 12347019

625.60 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10553283

  • 920.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Includes: Each unit contains one vial of reagent.
This item is not returnable. View return policy

Description

Description

293fectin™ Transfection Reagent is a proprietary, cationic lipid-based formulation for transfecting DNA into eukaryotic cells. This reagent is optimized for transfecting suspension 293 human embryonic kidney cells (e.g., FreeStyle™ 293-F cells, Cat. No. R790-07) in defined, serum-free FreeStyle™ 293 Expression Medium (12338-018), and is intended for use with the FreeStyle™ 293 Expression System (K9000-01).

293fectin™ Transfection Reagent provides the following advantages:

  • Demonstrates high transfection efficiency in suspension 293 cells and is also suitable for transfecting adherent 293 cells
  • Suspension FreeStyle™ 293-F cells may be transfected in FreeStyle™ 293 Expression Medium; no medium change is required
  • Add 293fectin™ reagent complexes directly to cells in culture medium
  • No need to remove complexes or change or add medium following transfection

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Each unit contains one vial of reagent. Store at 4°C.
293fectin™ is guaranteed stable for 6 months when stored properly.
Plasmid DNA
Transfection
293fectin™
1 mL
Established Cell Lines
Lipid-based Transfection
Not High-throughput Compatible (Manual)
Transfection Reagent
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.