missing translation for 'onlineSavingsMsg'
Learn More

Econix™ Bio-bins™ Tiger Stripe Paper Based Non-Sharps Offensive Waste Containers for Deep Landfill / Energy from Waste Only
GREENER_CHOICE

Use Econix™ Bio-bins™ The Bio-bin™ is a Paper based Non-sharps Container used to dispose of single use implements, tips, petri dishes, pads and other lab waste (excluding needles and blades) and reduce over filling issues for your non-sharp clinical waste. The Bio-bin has a built-in temporary seal, absorbent mat and is waterproof.

Brand:  Econix™ BEXBB2TS

 View more versions of this product

Product Code. 12983655

  • 189.00 € / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Description

Description

  • 96% paper based non-plastic single-use non-sharps waste container
  • Comes fold-flat for easy storage
  • Rigid, leakproof construction
  • Leakproof, built-in lid
  • Built-in absorbent Mat
  • UN 3291 approved
Includes temporary seal Autoclavable
Specifications

Specifications

2 L
Temporary and Permanent
Tiger Stripe
160 mm
Non-sterile
Autoclavable
Cardboard
UN3291
Space efficient fold-flat
260 mm
100 Pk.
160 mm
Offensive Clinical Waste
Non-Sharps Waste Container
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.