missing translation for 'onlineSavingsMsg'
Learn More

(2R,3R)-2-(Boc-amino)-3-benzyloxy-1-butanol, 97%, Thermo Scientific™

Product Code. 15449905
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133565-43-2
Molecular Formula C16H25NO4
Molecular Weight (g/mol) 295.38
MDL Number MFCD00235948
InChI Key RXDBGDZAKNELGW-UHFFFAOYNA-N
Synonym tert-butyl 2r,3r-3-benzyloxy-1-hydroxybutan-2-yl carbamate, boc-thr bzl-ol, boc-threoninol bzl, 2r,3r-2-boc-amino-3-benzyloxy-1-butanol, tert-butyl n-2r,3r-3-benzyloxy-1-hydroxybutan-2-yl carbamate, ambotzbal1043, n-boc-l-thr bzl-ol, n-boc-o-benzyl-l-threoninol, n-boc-2r,3r-2-amino-3-benzyloxy-1-butanol
PubChem CID 11109205
IUPAC Name tert-butyl N-[(2R,3R)-1-hydroxy-3-phenylmethoxybutan-2-yl]carbamate
SMILES CC(OCC1=CC=CC=C1)C(CO)NC(=O)OC(C)(C)C

Specifications

Quantity 250 mg
Formula Weight 295.38
Percent Purity 97%
Chemical Name or Material (2R,3R)-2-(Boc-amino)-3-benzyloxy-1-butanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'