missing translation for 'onlineSavingsMsg'
Learn More

(2R,3R)-2-(Fmoc-amino)-3-tert-butoxy-1-butanol, 97%, Thermo Scientific™

Product Code. 15409795
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 189337-28-8
Molecular Formula C23H29NO4
Molecular Weight (g/mol) 383.488
MDL Number MFCD00235950
InChI Key LBVPBNDGSCZOTB-QVKFZJNVSA-N
Synonym fmoc-thr tbu-ol, fmoc-threoninol tbu, 9h-fluoren-9-yl methyl 2r,3r-3-tert-butoxy-1-hydroxybutan-2-yl carbamate, 2r,3r-2-fmoc-amino-3-tert-butoxy-1-butanol, n-fmoc-l-thr tbu-ol, n-fmoc-2r,3r-2-amino-3-t-butoxy-1-butanol, 9h-fluoren-9-ylmethyl n-2r,3r-3-tert-butoxy-1-hydroxybutan-2-yl carbamate, ambotzfal1034, fmoc-l-threoninol tbu, fmoc-o-t-butyl-l-threoninol
PubChem CID 11143467
IUPAC Name 9H-fluoren-9-ylmethyl N-[(2R,3R)-1-hydroxy-3-[(2-methylpropan-2-yl)oxy]butan-2-yl]carbamate
SMILES CC(C(CO)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)OC(C)(C)C

Specifications

Quantity 250 mg
Formula Weight 383.49
Percent Purity 97%
Chemical Name or Material (2R,3R)-2-(Fmoc-amino)-3-tert-butoxy-1-butanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'