missing translation for 'onlineSavingsMsg'
Learn More
(2R,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane, 98%, Thermo Scientific™ Product Code.: 10284784

(2R,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane, 98%, Thermo Scientific™

Product Code. 10284784
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10284784

Brand: Thermo Scientific Acros 395010010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 98760-08-8
Molecular Formula C15H21NO3
Molecular Weight (g/mol) 263.34
MDL Number MFCD00671705,MFCD02258997
InChI Key NVPOUMXZERMIJK-UHFFFAOYNA-N
Synonym 2r,3s-3-tert-butoxycarbonyl amino-1,2-epoxy-4-phenylbutane, 2r,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, tert-butyl s-1-r-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl 1s,2r-oxiranyl-2-phenylethyl carbamate, threo-n-boc-l-phenylalanine epoxide, tert-butyl n-1s-1-2r-oxiran-2-yl-2-phenylethyl carbamate, 2r,3s-3-n-boc-amino-1-oxirane-4-phenylbutane, 2r,3s-3-tert-boc amino-1,2-epoxy-4-phenylbutane, atazanavir impurity c, pubchem5823
PubChem CID 9813904
IUPAC Name tert-butyl N-[(1S)-1-[(2R)-oxiran-2-yl]-2-phenylethyl]carbamate
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C1CO1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: <0.1%
Formula Weight 263.34
Percent Purity 98%
Chemical Name or Material (2R, 3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane
Product Identifier
  • (2R,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(2R,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.