missing translation for 'onlineSavingsMsg'
Learn More

3-(1,3-Oxazol-5-yl)benzoic acid, ≥97%, Thermo Scientific™

Product Code. 10094562
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 252928-82-8
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD06797481
InChI Key GDGXRJDVOKNSCX-UHFFFAOYSA-N
Synonym 3-1,3-oxazol-5-yl benzoic acid, 3-oxazol-5-yl benzoic acid, 3-5-oxazolyl benzoic acid, benzoic acid, 3-5-oxazolyl, 3-5-oxazolyl benzoicacid, benzoicacid, 3-5-oxazolyl
PubChem CID 7127813
IUPAC Name 3-(1,3-oxazol-5-yl)benzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C2=CN=CO2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 189.17
Percent Purity 97%
Chemical Name or Material 3-(1, 3-Oxazol-5-yl)benzoic acid
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 3-(1,3-Oxazol-5-yl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'