missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1-Methyl-1H-pyrazol-3-yl)benzaldehyde, 90%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 89.5 |
CAS Max % | 100.0 |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 3-1-methyl-1h-pyrazol-3-yl benzaldehyde, 3-1-methylpyrazol-3-yl benzaldehyde, benzaldehyde,3-1-methyl-1h-pyrazol-3-yl, benzaldehyde, 3-1-methyl-1h-pyrazol-3-yl, 3-3-formylphenyl-1-methyl-1h-pyrazole |
Formula Weight | 186.21 |
Percent Purity | 90% |
Chemical Name or Material | 3-(1-Methyl-1H-pyrazol-3-yl)benzaldehyde |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction