missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Naphthyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 15441039
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 881913-20-8
Molecular Formula C16H13BO2
Molecular Weight (g/mol) 248.088
MDL Number MFCD11045054
InChI Key HFXYUCJLQZCNPD-UHFFFAOYSA-N
Synonym 3-naphthalen-1-yl phenyl boronic acid, 3-naphthalen-1-yl phenylboronic acid, 3-1-naphthyl phenylboronic acid, 3-1-naphthyl benzeneboronic acid, boronic acid, 3-1-naphthalenyl phenyl, 3-1-naphthyl phenyl boronic acid, 3-1-naphthalenyl phenyl-boronic acid
PubChem CID 58881077
IUPAC Name (3-naphthalen-1-ylphenyl)boronic acid
SMILES B(C1=CC(=CC=C1)C2=CC=CC3=CC=CC=C32)(O)O

Specifications

Color White
Quantity 250 mg
Beilstein 18694809
Formula Weight 248.09
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-(1-Naphthyl)benzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'