missing translation for 'onlineSavingsMsg'
Learn More
3-(1-Pyrrolyl)benzoic acid, 97%, Thermo Scientific Chemicals Product Code.: 15455917

3-(1-Pyrrolyl)benzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 15455917
1 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy

Product Code. 15455917

Brand: Thermo Scientific Alfa Aesar H33124.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61471-45-2
Molecular Formula C11H8NO2
Molecular Weight (g/mol) 186.19
MDL Number MFCD02656610
InChI Key PODFNQCZFHLJPH-UHFFFAOYSA-M
Synonym 3-1h-pyrrol-1-yl benzoic acid, 3-pyrrol-1-yl-benzoic acid, benzoic acid, 3-1h-pyrrol-1-yl, n-3-carboxy phenylpyrrole, 3-1-pyrrolyl benzoic acid, 3-pyrrol-1-yl benzoic acid, 3-pyrrolylbenzoic acid, 1-3-carboxyphenyl-1h-pyrrole, benzoicacid,3-1h-pyrrol-1-yl, benzoicacid, 3-1h-pyrrol-1-yl
PubChem CID 736537
IUPAC Name 3-pyrrol-1-ylbenzoic acid
SMILES [O-]C(=O)C1=CC=CC(=C1)N1C=CC=C1

Specifications

Melting Point 175°C to 178°C
Quantity 1 g
Formula Weight 187.2
Percent Purity 97%
Chemical Name or Material 3-(1-Pyrrolyl)benzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(1-Pyrrolyl)benzoic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.