missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Imidazol-1-ylmethyl)benzoic acid hydrochloride, Tech., Thermo Scientific™

Product Code. 10306624
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 218131-32-9
Molecular Formula C11H11ClN2O2
Molecular Weight (g/mol) 238.671
MDL Number MFCD08690248
InChI Key YTCNDYWVEAFCMT-UHFFFAOYSA-N
Synonym 3-1h-imidazol-1-ylmethyl benzoic acid hydrochloride, 3-imidazol-1-ylmethyl-benzoic acid hydrochloride, 3-imidazol-1-ylmethyl benzoic acid hydrochloride, 3-imidazolylmethyl benzoic acid, chloride, 3-1h-imidazol-1-yl methyl benzoic acid hydrochloride, 3-1h-imidazol-1-yl methyl benzoic acid-hydrogen chloride 1/1
PubChem CID 18525763
IUPAC Name 3-(imidazol-1-ylmethyl)benzoic acid;hydrochloride
SMILES C1=CC(=CC(=C1)CN2C=CN=C2)C(=O)O.Cl

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 238.67
Percent Purity 90%
Grade Technical
Chemical Name or Material 3-(1H-Imidazol-1-ylmethyl)benzoic acid hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(1H-Imidazol-1-ylmethyl)benzoic acid hydrochloride, Tech., Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.