missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrazol-1-ylmethyl)aniline, 97%, Thermo Scientific™

Product Code. 10662475
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 892502-09-9
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.219
InChI Key JGTYTUGTENJXBY-UHFFFAOYSA-N
Synonym 3-1h-pyrazol-1-ylmethyl aniline, 3-1h-pyrazol-1-yl methyl aniline, 3-pyrazol-1-ylmethyl-phenylamine, 3-pyrazol-1-ylmethyl aniline, 3-1-pyrazolyl methyl aniline, 3-1h-pyrazol-1-ylmethyl phenyl amine, 3-pyrazolylmethyl phenylamine
PubChem CID 6485349
IUPAC Name 3-(pyrazol-1-ylmethyl)aniline
SMILES C1=CC(=CC(=C1)N)CN2C=CC=N2

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 173.22
Percent Purity 97%
Chemical Name or Material 3-(1H-Pyrazol-1-ylmethyl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'