missing translation for 'onlineSavingsMsg'
Learn More
3-(1H-Pyrrol-1-yl)benzene-1-carbothioamide, 97%, Thermo Scientific™ Product Code.: 10029471

3-(1H-Pyrrol-1-yl)benzene-1-carbothioamide, 97%, Thermo Scientific™

Product Code. 10029471
1 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy

Product Code. 10029471

Brand: Thermo Scientific Maybridge SPB01074DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175276-79-6
Molecular Formula C11H10N2S
Molecular Weight (g/mol) 202.275
MDL Number MFCD00052522
InChI Key LKZUYCBQOPNMNQ-UHFFFAOYSA-N
Synonym 3-1h-pyrrol-1-yl benzene-1-carbothioamide, 3-pyrrol-1-yl benzenecarbothioamide, 3-1h-pyrrol-1-yl benzenecarbothioamide, benzenecarbothioamide,3-1h-pyrrol-1-yl, 3-1h-pyrrol-1-yl benzothioamide, amino 3-pyrrolylphenyl methane-1-thione
PubChem CID 2799009
IUPAC Name 3-pyrrol-1-ylbenzenecarbothioamide
SMILES C1=CN(C=C1)C2=CC=CC(=C2)C(=S)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 202.28
Percent Purity 97%
Chemical Name or Material 3-(1H-pyrrol-1-yl)benzene-1-carbothioamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(1H-Pyrrol-1-yl)benzene-1-carbothioamide, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.