missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Tetrazol-5-yl)phenol, 97%, Thermo Scientific Chemicals

Product Code. 11491800
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96859-34-6
Molecular Formula C7H6N4O
Molecular Weight (g/mol) 162.15
MDL Number MFCD06797247
InChI Key IZORRBUQWFSCII-UHFFFAOYSA-N
Synonym 3-1h-tetrazol-5-yl phenol, 5-3-hydroxyphenyl tetrazole, 3-2h-tetrazol-5-yl phenol, 3-2h-1,2,3,4-tetrazol-5-yl phenol, 3-1h-1,2,3,4-tetrazol-5-yl phenol, m-tetrazolylphenol, 3-2h-tetrazol-5-yl-phenol, 5-3-hydroxyphenyl-1h-tetrazole
PubChem CID 13455763
SMILES OC1=CC=CC(=C1)C1=NNN=N1

Specifications

Melting Point ∽220°C (decomposition)
Quantity 5 g
IUPAC Name 3-(2H-tetrazol-5-yl)phenol
Formula Weight 162.16
Percent Purity 97%
Chemical Name or Material 3-(1H-Tetrazol-5-yl)phenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'