Learn More
3-(2-Aminoethylamino)propylmethyldimethoxysilane, 97%, Thermo Scientific Chemicals
23.82 € - 151.00 €
Chemical Identifiers
CAS | 3069-29-2 |
---|---|
Molecular Formula | C8H22N2O2Si |
Molecular Weight (g/mol) | 206.36 |
MDL Number | MFCD00054776 |
InChI Key | MQWFLKHKWJMCEN-UHFFFAOYSA-N |
Synonym | n-2-aminoethyl-3-aminopropylmethyldimethoxysilane, 3-2-aminoethylamino propyl-dimethoxymethylsilane, 3-2-aminoethylamino propyldimethoxymethylsilane, 3-2-aminoethylamino propylmethyldimethoxysilane, n-3-dimethoxymethylsilyl propyl ethylenediamine, 1,2-ethanediamine, n-3-dimethoxymethylsilyl propyl, n-3-dimethoxy methyl silyl propyl ethane-1,2-diamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 1,2-ethanediamine, n1-3-dimethoxymethylsilyl propyl |
PubChem CID | 76482 |
IUPAC Name | N'-[3-[dimethoxy(methyl)silyl]propyl]ethane-1,2-diamine |
SMILES | CO[Si](C)(CCCNCCN)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11416396
|
Thermo Scientific Alfa Aesar
B23159.14 |
25 g |
23.82 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11426396
|
Thermo Scientific Alfa Aesar
B23159.22 |
100 g |
48.24 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11436396
|
Thermo Scientific Alfa Aesar
B23159.36 |
500 g |
151.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3069-29-2 | |
206.36 | |
MQWFLKHKWJMCEN-UHFFFAOYSA-N | |
76482 | |
CO[Si](C)(CCCNCCN)OC |
C8H22N2O2Si | |
MFCD00054776 | |
n-2-aminoethyl-3-aminopropylmethyldimethoxysilane, 3-2-aminoethylamino propyl-dimethoxymethylsilane, 3-2-aminoethylamino propyldimethoxymethylsilane, 3-2-aminoethylamino propylmethyldimethoxysilane, n-3-dimethoxymethylsilyl propyl ethylenediamine, 1,2-ethanediamine, n-3-dimethoxymethylsilyl propyl, n-3-dimethoxy methyl silyl propyl ethane-1,2-diamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 1,2-ethanediamine, n1-3-dimethoxymethylsilyl propyl | |
N'-[3-[dimethoxy(methyl)silyl]propyl]ethane-1,2-diamine |
Specifications
265°C | |
C8H22N2O2Si | |
UN2735 | |
90°C (194°F) | |
MQWFLKHKWJMCEN-UHFFFAOYSA-N | |
N'-[3-[dimethoxy(methyl)silyl]propyl]ethane-1,2-diamine | |
76482 | |
97% | |
1.449 | |
0.968 |
3069-29-2 | |
MFCD00054776 | |
2074259 | |
n-2-aminoethyl-3-aminopropylmethyldimethoxysilane, 3-2-aminoethylamino propyl-dimethoxymethylsilane, 3-2-aminoethylamino propyldimethoxymethylsilane, 3-2-aminoethylamino propylmethyldimethoxysilane, n-3-dimethoxymethylsilyl propyl ethylenediamine, 1,2-ethanediamine, n-3-dimethoxymethylsilyl propyl, n-3-dimethoxy methyl silyl propyl ethane-1,2-diamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 1,2-ethanediamine, n1-3-dimethoxymethylsilyl propyl | |
CO[Si](C)(CCCNCCN)OC | |
206.36 | |
206.36 | |
Amine-like | |
Air and moisture sensitive | |
3-(2-Aminoethylamino)propylmethyldimethoxysilane |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P362-P370+P378q-P501c
H227-H302-H315-H317-H318
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 221-336-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only