Learn More
3-(2-Aminoethylamino)propyltrimethoxysilane, 96%, Thermo Scientific Chemicals
30.40 € - 178.00 €
Chemical Identifiers
CAS | 1760-24-3 |
---|---|
Molecular Formula | C8H22N2O3Si |
Molecular Weight (g/mol) | 222.36 |
MDL Number | MFCD00008173 |
InChI Key | PHQOGHDTIVQXHL-UHFFFAOYSA-N |
Synonym | n-3-trimethoxysilyl propyl ethylenediamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 3-2-aminoethylamino propyltrimethoxysilane, en-aptas, silicone a-1120, prosil 3128, aas-m, n1-3-trimethoxysilyl propyl ethane-1,2-diamine, unii-28zcs5ga8g, 3-2-aminoethyl aminopropyl trimethoxysilane |
PubChem CID | 15659 |
IUPAC Name | N'-(3-trimethoxysilylpropyl)ethane-1,2-diamine |
SMILES | CO[Si](CCCNCCN)(OC)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11358925
|
Thermo Scientific Alfa Aesar
L14043.14 |
25 g |
30.40 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11368925
|
Thermo Scientific Alfa Aesar
L14043.22 |
100 g |
54.70 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11378925
|
Thermo Scientific Alfa Aesar
L14043.36 |
500 g |
178.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-(2-Aminoethylamino) propyltrimethoxysilane acts as a silane coupling agent. It is useful for chemical research. It is also employed as an adhesives and sealant chemical. Further, it is used as a paint additive and coating additive. In addition to this, it plays an important role in the production of petroleum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(2-Aminoethylamino) propyltrimethoxysilane acts as a silane coupling agent. It is useful for chemical research. It is also employed as an adhesives and sealant chemical. Further, it is used as a paint additive and coating additive. In addition to this, it plays an important role in the production of petroleum.
Solubility
Miscible with toluene.
Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and water.
Chemical Identifiers
1760-24-3 | |
222.36 | |
PHQOGHDTIVQXHL-UHFFFAOYSA-N | |
15659 | |
CO[Si](CCCNCCN)(OC)OC |
C8H22N2O3Si | |
MFCD00008173 | |
n-3-trimethoxysilyl propyl ethylenediamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 3-2-aminoethylamino propyltrimethoxysilane, en-aptas, silicone a-1120, prosil 3128, aas-m, n1-3-trimethoxysilyl propyl ethane-1,2-diamine, unii-28zcs5ga8g, 3-2-aminoethyl aminopropyl trimethoxysilane | |
N'-(3-trimethoxysilylpropyl)ethane-1,2-diamine |
Specifications
261°C to 263°C | |
C8H22N2O3Si | |
UN2735 | |
136°C (276°F) | |
Miscible with toluene. | |
CO[Si](CCCNCCN)(OC)OC | |
222.36 | |
222.36 | |
Ammonia-like | |
Air and moisture sensitive | |
3-(2-Aminoethylamino)propyltrimethoxysilane |
1760-24-3 | |
MFCD00008173 | |
636230 | |
n-3-trimethoxysilyl propyl ethylenediamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 3-2-aminoethylamino propyltrimethoxysilane, en-aptas, silicone a-1120, prosil 3128, aas-m, n1-3-trimethoxysilyl propyl ethane-1,2-diamine, unii-28zcs5ga8g, 3-2-aminoethyl aminopropyl trimethoxysilane | |
PHQOGHDTIVQXHL-UHFFFAOYSA-N | |
N'-(3-trimethoxysilylpropyl)ethane-1,2-diamine | |
15659 | |
96% | |
1.4435 | |
1.01 |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P362-P501c
H315-H317-H318
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 217-164-6
missing translation for 'rtecsNumber' : KV7400000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only