missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Furyl)-1-methyl-1H-pyrazole-5-carboxylic acid, ≥97%, Thermo Scientific™

Product Code. 10507854
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 859851-00-6
Molecular Formula C9H8N2O3
Molecular Weight (g/mol) 192.17
MDL Number MFCD08271950
InChI Key MOPCEJWYTICWJM-UHFFFAOYSA-N
Synonym 3-2-furyl-1-methyl-1h-pyrazole-5-carboxylic acid, 5-furan-2-yl-2-methylpyrazole-3-carboxylic acid, 3-furan-2-yl-1-methyl-1h-pyrazole-5-carboxylic acid, 1h-pyrazole-5-carboxylicacid, 3-2-furanyl-1-methyl, 3-fur-2-yl-1-methyl-1h-pyrazole-5-carboxylic acid, 3-2-furyl-1-methylpyrazole-5-carboxylic acid
PubChem CID 7537643
SMILES CN1N=C(C=C1C(O)=O)C1=CC=CO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 5-(furan-2-yl)-2-methylpyrazole-3-carboxylic acid
Formula Weight 192.17
Percent Purity 97%
Chemical Name or Material 3-(2-Furyl)-1-methyl-1H-pyrazole-5-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'