missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Furyl)aniline, 97%, Thermo Scientific™

Product Code. 10586314
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 102269-42-1
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD04039071
InChI Key SLNKACMTMZYMNA-UHFFFAOYSA-N
Synonym 3-furan-2-yl aniline, 3-2-furyl aniline, benzenamine,3-2-furanyl, acmc-1c42n, 3-2-furyl aniline hydrochloride
PubChem CID 4161309
IUPAC Name 3-(furan-2-yl)aniline
SMILES C1=CC(=CC(=C1)N)C2=CC=CO2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 3-(2-Furyl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'