missing translation for 'onlineSavingsMsg'
Learn More

3,3,3-Trifluoro-1-propanol, 97%, Thermo Scientific Chemicals

Product Code. 11323816
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3,3,3-Trifluoro-1-propanol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3,3-Trifluoro-1-propanol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2240-88-2
Molecular Formula C3H5F3O
Molecular Weight (g/mol) 114.067
MDL Number MFCD00190649
InChI Key HDBGBTNNPRCVND-UHFFFAOYSA-N
Synonym 3,3,3-trifluoro-1-propanol, 3,3,3-trifluoropropanol, 1-propanol, 3,3,3-trifluoro, trifluoropropanol, acmc-1cqb1, 3,3,3-trifluoropropanol-1, 2-trifluoromethyl ethan-1-ol, 3,3,3-trifluoro-propan-1-ol, poly difluoromethylene , alpha-fluoro-omega-2-hydroxyethyl, 3-hydroxy-1,1,1-trifluoropropane
PubChem CID 137512
IUPAC Name 3,3,3-trifluoropropan-1-ol
SMILES C(CO)C(F)(F)F

Specifications

Density 1.294
Boiling Point 100°C to 101°C
Flash Point 37°C (98°F)
Refractive Index 1.32
Quantity 1 g
UN Number UN1987
Beilstein 1734777
Solubility Information Fully miscible in water.
Formula Weight 114.07
Percent Purity 97%
Chemical Name or Material 3,3,3-Trifluoro-1-propanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'