Learn More
3-(3,4-Dichlorophenyl)-1,1-dimethylurea, 97%, Thermo Scientific Chemicals
Description
A phenyl urea pre-emergent agrochemical. Also an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyestuff
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
A phenyl urea pre-emergent agrochemical. Also an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyestuff
Solubility
Soluble in water (0.049g/L), chloroform and methanol. Very slightly soluble in many organic solvents.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Specifications
Specifications
Melting Point | 157°C to 160°C |
Density | 1.48 |
Boiling Point | 180°C to 190°C (decomposition) |
Odor | Odorless |
Quantity | 250 g |
UN Number | UN3077 |
Beilstein | 2215168 |
Merck Index | 14,3382 |
Solubility Information | Soluble in water (0.049g/L),chloroform and methanol. Very slightly soluble in many organic solvents. |
Formula Weight | 233.1 |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.