missing translation for 'onlineSavingsMsg'
Learn More
3-(3,4-Dihydroxyphenyl)-L-alanine, 99%, Thermo Scientific Chemicals Product Code.: 10606762

3-(3,4-Dihydroxyphenyl)-L-alanine, 99%, Thermo Scientific Chemicals

Product Code. 10606762
5 g, Glass Bottle
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10606762

Brand: Thermo Scientific Acros 167530050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-(3,4-Dihydroxyphenyl)-L-alanine (levodopa orL-dopa), CAS # 59-92-7, is a dopamine precursor molecule.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• 3-(3,4-Dihydroxyphenyl)-L-alanine, or L-dopa, is converted to dopamine by DOPA decarboxylase in the body

• Upon conversion to dopamine, it acts as an excitatory neurotransmitter

Applications

• L-dopa is useful in the laboratory study of Parkinson’s to replenish dopamine in depleted circuits

• L-dopa has also been used for laboratory research on hypertension and melanocyte activity

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-92-7
Molecular Formula C9H11NO4
Molecular Weight (g/mol) 197.19
MDL Number MFCD00002598
InChI Key WTDRDQBEARUVNC-LURJTMIESA-N
Synonym levodopa, l-dopa, dopar, 3-hydroxy-l-tyrosine, 3,4-dihydroxy-l-phenylalanine, larodopa, bendopa, levopa, dihydroxy-l-phenylalanine, cidandopaShow MoreShow Less
PubChem CID 6047
ChEBI CHEBI:15765
IUPAC Name (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C(O)=C1)C(O)=O

Specifications

Color Cream to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Loss on Drying 1% max.
Packaging Glass Bottle
Linear Formula (HO)2C6H3CH2CH(NH2)CO2H
Beilstein 14, IV, 2492
Merck Index 15, 5516
Sulfated Ash 0.1% max.
Quantity 5 g
Formula Weight 197.19
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 3-(3, 4-Dihydroxyphenyl)-L-alanine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0455700 Certificate TypeCertificate of Analysis Date25/06/2024 Catalog Number167530250,167531000,167530050
Lot NumberA0450216 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number167530250,167531000,167530050
Lot NumberA0395862 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number167530250,167531000,167535000,167530050
Lot NumberA0396107 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number167530250,167531000,167535000,167530050
Lot NumberA0397062 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number167530250,167531000,167535000,167530050
compliance-icons
Product Identifier
  • L-3-(3,4-Dihydroxyphenyl)alanine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : L-3-(3,4-Dihydroxyphenyl)alanine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(3,4-Dihydroxyphenyl)-L-alanine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.