missing translation for 'onlineSavingsMsg'
Learn More

3-(3,4-Dimethoxyphenyl)propionic acid, 98%, Thermo Scientific Chemicals

Product Code. 11436877
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2107-70-2
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.229
MDL Number MFCD00002774
InChI Key LHHKQWQTBCTDQM-UHFFFAOYSA-N
Synonym 3-3,4-dimethoxyphenyl propionic acid, 3-3,4-dimethoxyphenyl propanoic acid, 3,4-dimethoxyhydrocinnamic acid, benzenepropanoic acid, 3,4-dimethoxy, 3-3,4-dimethoxyphenyl-propionic acid, 3-3,4-dimethoxy-phenyl-propionic acid, unii-6xo32zsp1d, 6xo32zsp1d, 3-3',4'-dimethoxyphenyl propanoic acid, 3-3,4-dimethoxyphenyl propionicacid
PubChem CID 75019
ChEBI CHEBI:44235
IUPAC Name 3-(3,4-dimethoxyphenyl)propanoic acid
SMILES COC1=C(C=C(C=C1)CCC(=O)O)OC

Specifications

Melting Point 96°C to 99°C
Quantity 5 g
Beilstein 2696272
Formula Weight 210.23
Percent Purity 98%
Chemical Name or Material 3-(3,4-Dimethoxyphenyl)propionic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'