missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetra-tert-butyldiphenoquinone, 98%, Thermo Scientific Chemicals

Product Code. 15294238
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2455-14-3
Molecular Formula C28H40O2
Molecular Weight (g/mol) 408.63
InChI Key GQIGHOCYKUBBOE-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetra-tert-butyldiphenoquinone, 3,3',5,5'-tetra-tert-butyl-4,4'-diphenoquinone, 3,3',5,5'-tetra-tert-butyl-1,1'-bi cyclohexylidene-2,2',5,5'-tetraene-4,4'-dione, unii-4m92z38q9a, 3,3',5,5'-tetra-tert-butyl-4,4'-dibenzoquinone, bi-2,5-cyclohexadien-1-ylidene-4,4'-dione, 3,3',5,5'-tetra-tert-butyl, 2,2',6,6'-tetra-tert-butyldiphenylquinone, 2,6-ditert-butyl-4-3,5-ditert-butyl-4-oxocyclohexa-2,5-dien-1-ylidene cyclohexa-2,5-dien-1-one, pubchem13798, rarechem bw gc 0005
PubChem CID 225283
IUPAC Name 2,6-ditert-butyl-4-(3,5-ditert-butyl-4-oxocyclohexa-2,5-dien-1-ylidene)cyclohexa-2,5-dien-1-one
SMILES CC(C)(C)C1=CC(=C2C=C(C(=O)C(=C2)C(C)(C)C)C(C)(C)C)C=C(C1=O)C(C)(C)C

Specifications

Melting Point 247.0°C to 248.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 2 g
Formula Weight 408.63
Percent Purity 98%
Chemical Name or Material 3,3',5,5'-Tetra-tert-butyldiphenoquinone
compliance-icons
Product Identifier
  • 3,3',5,5'-Tetra-tert-butyldiphenoquinone, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.