missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetramethylbenzidine, 98%, Thermo Scientific Chemicals

Product Code. 11451548
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3,3',5,5'-Tetramethylbenzidine, 98%, or TMB, CAS # 54827-17-7, is a colorimetric reagent used in laboratory-based substrate detection.

Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes; particularly useful in ELISA techniques, blue result can be read spectrophotometrically at 370 or 650 nm3,3',5,5'-Tetramethylbenzidine is used as a clinical reagent for testing blood. It is also a chromogenic substrate which is used in staining procedures in immunohistochemistry. Further, it is used as a visualizing reagent in enzyme-linked immunosorbent assays. In addition to this, it is used as a replacement for carcinogenic compounds such as benzidine and o-phenylenediamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• 3,3',5,5'-Tetramethylbenzidine, also known as 4,4'-Diamino-3,3',5,5'-tetramethylbiphenyl or TMB, is a colorimetric substrate for detection of horseradish peroxidase-labeled probes
• Oxidation of 3,3',5,5'-Tetramethylbenzidine results in a pale blue end product that can be detected at 370 or 655 nm. Sulfuric acid (H2SO4) is used to stop the oxidation reaction and stabilize the reading, resulting in a yellow color that can be read at 450 nm
Application
• 3,3',5,5'-Tetramethylbenzidine is used as a chromogenic visualization agent in enzyme-linked immunosorbent assays (ELISA) or immunohistochemistry (IHC) techniques
• 3,3',5,5'-Tetramethylbenzidine has been used to detect molecules in aqueous samples such as copper ion and hypochlorite
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54827-17-7
Molecular Formula C16H20N2
Molecular Weight (g/mol) 240.35
MDL Number MFCD00007748
InChI Key UAIUNKRWKOVEES-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetramethylbenzidine, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 3,5,3',5'-tetramethylbenzidine, tmb, 3,3',5,5'-tetramethyl benzidine, tmb substrate, bm blue, ccris 4727, tmb plus, tmb, insoluble
PubChem CID 41206
IUPAC Name 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline
SMILES CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C

Specifications

Melting Point 168°C to 171°C
Quantity 25 g
Beilstein 2808541
Sensitivity Light and moisture sensitive
Solubility Information Soluble in ethyl acetate,ethanol,acetone,chloroform,toluene and dimethyl sulfoxide. Insoluble in water.
Formula Weight 240.35
Percent Purity 98%
Chemical Name or Material 3,3',5,5'-Tetramethylbenzidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'