missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetramethylbenzidine dihydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 15495586
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64285-73-0
Molecular Formula C16H22Cl2N2
Molecular Weight (g/mol) 313.27
MDL Number MFCD00012961
InChI Key NYNRGZULARUZCC-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetramethylbenzidine dihydrochloride, tmb dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3',5,5'-tetramethyl-, dihydrochloride, tetramethylbenzidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3',5,5'-tetramethyl-, hydrochloride 1:2, 4-4-amino-3,5-dimethylphenyl-2,6-dimethylaniline dihydrochloride, 3,3',5,5'-tetramethylbenzidine dihydrochloride hydrate, 3,3',5,5'-tetramethyl 1,1'-biphenyl-4,4'-diamine dihydrochloride
PubChem CID 174033
IUPAC Name 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].CC1=CC(=CC(C)=C1N)C1=CC(C)=C(N)C(C)=C1

Specifications

Melting Point 300°C
Quantity 1 g
Sensitivity Light and moisture sensitive
Formula Weight 313.27
Percent Purity 99%
Chemical Name or Material 3,3',5,5'-Tetramethylbenzidine dihydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'