missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate, 98+%, Thermo Scientific Chemicals

Product Code. 10318220
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 207738-08-7, 54827-17-7
CAS Min % 98.0
CAS Max % 100.0
Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98% min. (on dry substance) (Argentometry)
Packaging Glass bottle
Molecular Formula C16H22Cl2N2
Linear Formula 2HCl·xH2O
MDL Number MFCD00150104
Quantity 1 g
Synonym 3,3',5,5'-tetramethylbenzidine dihydrochloride dihydrate, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride dihydrate, 3,3',5,5'-tetramethylbenzidine dihydrochloride hydrate, pubchem9044, tetramethylbenzidine dihydrate dihydrochloride
Solubility Information Solubility in water: moderately soluble. Other solubilities: soluble in methanol
InChI Key NYNRGZULARUZCC-UHFFFAOYSA-N
SMILES Cl.Cl.CC1=CC(=CC(C)=C1N)C1=CC(C)=C(N)C(C)=C1
IUPAC Name 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline;dihydrate;dihydrochloride
Molecular Weight (g/mol) 313.27
PubChem CID 19836601
Formula Weight 313.26
Percent Purity 98+%
Physical Form Fluffy Powder
Chemical Name or Material 3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate, 98+%
Show More Show Less
Product Identifier
  • 3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'