missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 3,3',5,5'-Tetramethylbenzidine Liquid Substrate

Product Code. 11442642 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 11442642

Brand: MP Biomedicals™ 0215234680

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

3,3',5,5'-Tetramethylbenzide Liquid Substrate is a chromogenic substrate for Horse Radish Peroxidase (HRP). TMB Solution is a convenient, non toxic and ready to use stable chromogen solution suitable for peroxidase and pseudo-peroxidase reactions in ELISA, it provides consistent performance and sensitivity to deliver reproducible results.

  • Ready to use 1.25 mmol/L stablized chromogen solution
  • Metabolism of the TMB substrate by HRP creates a soluble, bluish reaction product that may be quantified by measuring absorbance at 370 or 665 nm.
  • Reaction can be stopped by addition of 1 volume of 1N hydrocloric or sulfurc acid to yield a yellow reaction product that can be measured by absorbance at 450 nm (within one hour).
TRUSTED_SUSTAINABILITY

Specifications

Product Type Liquid Substrate
Format Solution
Quantity 100 mL
Storage Requirements 4°C, protect from light

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.