missing translation for 'onlineSavingsMsg'
Learn More

3-[3-(Benzyloxy)phenyl]propionic acid, 96%, Thermo Scientific Chemicals

Product Code. 15426787
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57668-34-5
Molecular Formula C16H16O3
Molecular Weight (g/mol) 256.301
MDL Number MFCD09258905
InChI Key HVXOYUOKPLHWNU-UHFFFAOYSA-N
Synonym 3-3-benzyloxy phenyl propanoic acid, benzenepropanoic acid, 3-phenylmethoxy, 3-3-benzyloxy-phenyl-propanoic acid, 3-3-benzyloxy phenyl propionic acid, 3-3-phenylmethyl oxy phenyl propanoic acid
PubChem CID 21938504
IUPAC Name 3-(3-phenylmethoxyphenyl)propanoic acid
SMILES C1=CC=C(C=C1)COC2=CC=CC(=C2)CCC(=O)O

Specifications

Melting Point 83°C to 85°C
Quantity 1 g
Formula Weight 256.3
Percent Purity 96%
Chemical Name or Material 3-[3-(Benzyloxy)phenyl]propionic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'